Learn More
Cayman Chemical Oxcarbazepine, Purity: ≥98%, CAS: 28721-07-5, 50 mg
SDP
Catalog No. 502966075
Encompass_Preferred
A prodrug form of 10,11-dihydro-10-hydroxy carbamazepine; inhibits high-frequency repetitive firing of rat spinal cord neurons at 1 µM; protects against electroshock-induced tonic hindlimb extension (ED50 = 10-20 mg/kg), as well as PTZ- or picrotoxin-induced seizures in rodents (ED50s = 23-30 and 150-250 mg/kg, respectively)
Catalog No. 50-296-6075 Supplier Cayman Chemical Supplier No. 1734050 MG
A prodrug form of 10,11-dihydro-10-hydroxy carbamazepine; inhibits high-frequency repetitive firing of rat spinal cord neurons at 1 µM; protects against electroshock-induced tonic hindlimb extension (ED50 = 10-20 mg/kg), as well as PTZ- or picrotoxin-induced seizures in rodents (ED50s = 23-30 and 150-250 mg/kg, respectively)