Learn More
Apexbio Technology LLC Oxcarbazepine 28721-07-5 10mg
SDP
Catalog No. 502240817
Encompass_Preferred
Oxcarbazepine (CAS 28721-07-5) is a small molecule that modulates voltage-gated sodium channels a critical target in neuronal excitability studies It impedes the association of [3H]batrachotoxin ([3H]BTX) with sodium channels demonstrating an IC50 of 160 M and restricts sodium ion influx as indicated by inhibition of 22Na uptake into rat brain synaptosomes with an IC50 of approximately 100 M These properties make oxcarbazepine valuable for investigating sodium channel function and regulation in neuropharmacology and for exploring mechanisms underlying neuronal signaling and excitotoxicity
This item is not returnable. View return policy
Catalog No. 50-224-0817 Supplier Apexbio Technology LLC Supplier No. B227910
Oxcarbazepine (CAS 28721-07-5) is a small molecule that modulates voltage-gated sodium channels a critical target in neuronal excitability studies It impedes the association of [3H]batrachotoxin ([3H]BTX) with sodium channels demonstrating an IC50 of 160 M and restricts sodium ion influx as indicated by inhibition of 22Na uptake into rat brain synaptosomes with an IC50 of approximately 100 M These properties make oxcarbazepine valuable for investigating sodium channel function and regulation in neuropharmacology and for exploring mechanisms underlying neuronal signaling and excitotoxicity