Learn More
Selleck Chemical LLC Oxcarbazepine S1391-10mg
SDP
Catalog No. 503110361
Encompass_Preferred
Oxcarbazepine (GP47680) inhibits the binding of [3H]BTX to sodium channels with IC50 of 160 M and also inhibits the influx of 22Na into rat brain synaptosomes with IC50 about 100 M
Catalog No. 50-311-0361 Supplier Selleck Chemical LLC Supplier No. S139110MG
Oxcarbazepine (GP47680) inhibits the binding of [3H]BTX to sodium channels with IC50 of 160 M and also inhibits the influx of 22Na into rat brain synaptosomes with IC50 about 100 M