Learn More
Apexbio Technology LLC Oxcarbazepine 28721-07-5 10mM (in 1mL DMSO)
SDP
Catalog No. 5000090824
Encompass_Preferred
Oxcarbazepine (CAS 28721-07-5) is a small molecule that modulates voltage-gated sodium channels a critical target in neuronal excitability studies It impedes the association of [3H]batrachotoxin ([3H]BTX) with sodium channels demonstrating an IC50 of 160 M and restricts sodium ion influx as indicated by inhibition of 22Na uptake into rat brain synaptosomes with an IC50 of approximately 100 M These properties make oxcarbazepine valuable for investigating sodium channel function and regulation in neuropharmacology and for exploring mechanisms underlying neuronal signaling and excitotoxicity
Catalog No. 50-000-90824 Supplier Apexbio Technology LLC Supplier No. B22795.1
Oxcarbazepine (CAS 28721-07-5) is a small molecule that modulates voltage-gated sodium channels a critical target in neuronal excitability studies It impedes the association of [3H]batrachotoxin ([3H]BTX) with sodium channels demonstrating an IC50 of 160 M and restricts sodium ion influx as indicated by inhibition of 22Na uptake into rat brain synaptosomes with an IC50 of approximately 100 M These properties make oxcarbazepine valuable for investigating sodium channel function and regulation in neuropharmacology and for exploring mechanisms underlying neuronal signaling and excitotoxicity