Learn More
Medchemexpress LLC Oxcarbazepine | 28721-07-5 | 99.9% | 252.27 | C15H12N2O2 | 10 MG
SDP
Catalog No. 5000298231
Encompass_Preferred
Oxcarbazepine (Standard) is an analytical reference standard of oxcarbazepine supplied for research and analytical applications. It is intended for qualitative, quantitative, and methodological experiments in HPLC, GC, and MS, and is commonly used for method development and assay calibration. CAS 28721-07-5.
  • Analytical reference standard for HPLC, GC, and MS.
  • High purity (99.88%), suitable for quantitative analysis.
  • Available in multiple small package sizes, including 10 mg.
  • Supplied with certificate of analysis and safety data sheet.
  • Intended for method development, assay calibration, and quality control.
Catalog No. 50-002-98231 Supplier Medchemexpress LLC Supplier No. HYB0114R10MG
Oxcarbazepine (Standard) is an analytical reference standard of oxcarbazepine supplied for research and analytical applications. It is intended for qualitative, quantitative, and methodological experiments in HPLC, GC, and MS, and is commonly used for method development and assay calibration. CAS 28721-07-5.
  • Analytical reference standard for HPLC, GC, and MS.
  • High purity (99.88%), suitable for quantitative analysis.
  • Available in multiple small package sizes, including 10 mg.
  • Supplied with certificate of analysis and safety data sheet.
  • Intended for method development, assay calibration, and quality control.