Learn More
Medchemexpress LLC Oxcarbazepine | 28721-07-5 | 99.9% | C15H12N2O2 | 50 MG
SDP
Catalog No. 5000432432
Encompass_Preferred
Oxcarbazepine is a sodium channel blocker that significantly inhibits glioblastoma cell growth and induces apoptosis or G2/M arrest in glioblastoma cell lines. It exhibits anti-cancer and anticonvulsant effects.
  • Sodium channel blocker
  • Significantly inhibits glioblastoma cell growth
  • Induces apoptosis or G2/M arrest in glioblastoma cell lines
  • Anti-cancer effects
  • Anticonvulsant effects
Catalog No. 50-004-32432 Supplier Medchemexpress LLC Supplier No. HYB011450MG
Oxcarbazepine is a sodium channel blocker that significantly inhibits glioblastoma cell growth and induces apoptosis or G2/M arrest in glioblastoma cell lines. It exhibits anti-cancer and anticonvulsant effects.
  • Sodium channel blocker
  • Significantly inhibits glioblastoma cell growth
  • Induces apoptosis or G2/M arrest in glioblastoma cell lines
  • Anti-cancer effects
  • Anticonvulsant effects