Learn More
Medchemexpress LLC Oxcarbazepine | 28721-07-5 | MFCD00865307 | 99.9% | C15H12N2O2 | 500 MG
SDP
Catalog No. 5000432062
Encompass_Preferred
Oxcarbazepine is a sodium channel blocker with anti-cancer and anticonvulsant effects. It significantly inhibits glioblastoma cell growth and induces apoptosis or G2/M arrest in glioblastoma cell lines.
  • Significantly inhibits glioblastoma cell growth
  • Induces apoptosis or G2/M arrest in glioblastoma cell lines
  • Exhibits anticonvulsant effects
Catalog No. 50-004-32062 Supplier Medchemexpress LLC Supplier No. HYB0114500MG
Oxcarbazepine is a sodium channel blocker with anti-cancer and anticonvulsant effects. It significantly inhibits glioblastoma cell growth and induces apoptosis or G2/M arrest in glioblastoma cell lines.
  • Significantly inhibits glioblastoma cell growth
  • Induces apoptosis or G2/M arrest in glioblastoma cell lines
  • Exhibits anticonvulsant effects