Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Oxcarbazepine-d4 | 1020719-71-4 | C15H8D4N2O2 | 2.5 MG
SDP

Catalog No. 5000433455
Encompass_Preferred

Oxcarbazepine-d4 is a deuterium-labeled version of Oxcarbazepine, which acts as a sodium channel blocker. It significantly inhibits glioblastoma cell growth, inducing apoptosis or G2/M arrest in glioblastoma cell lines. This compound exhibits both anti-cancer and anticonvulsant effects.

  • Deuterium labeled oxcarbazepine
  • Acts as a sodium channel blocker
  • Molecular weight: 256.29
  • Formula: C15H8D4N2O2
  • Can be utilized as a tracer
  • Can serve as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Incorporated into drug molecules for quantitation during drug development
  • Potential to influence pharmacokinetic and metabolic profiles of drugs

Catalog No. 50-004-33455 Supplier Medchemexpress LLC Supplier No. HYB0114S2.5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Oxcarbazepine-d4 is a deuterium-labeled version of Oxcarbazepine, which acts as a sodium channel blocker. It significantly inhibits glioblastoma cell growth, inducing apoptosis or G2/M arrest in glioblastoma cell lines. This compound exhibits both anti-cancer and anticonvulsant effects.

  • Deuterium labeled oxcarbazepine
  • Acts as a sodium channel blocker
  • Molecular weight: 256.29
  • Formula: C15H8D4N2O2
  • Can be utilized as a tracer
  • Can serve as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Incorporated into drug molecules for quantitation during drug development
  • Potential to influence pharmacokinetic and metabolic profiles of drugs

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.