Learn More
Medchemexpress LLC Oxcarbazepine (Standard) | 28721-07-5 | 99.9% | C₁₅H₁₂N₂O₂ | 100 MG
SDP
Catalog No. 5000432918
Encompass_Preferred
Oxcarbazepine (Standard) is the analytical standard of Oxcarbazepine, intended for research and analytical applications. It is a sodium channel blocker that significantly inhibits glioblastoma cell growth and induces apoptosis or G2/M arrest in glioblastoma cell lines. It exhibits anti-cancer and anticonvulsant effects. This product is for research use only.
  • Analytical standard
  • Used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS
  • Sodium channel blocker
  • Inhibits glioblastoma cell growth
  • Induces apoptosis or G2/M arrest in glioblastoma cell lines
  • Anti-cancer effects
  • Anticonvulsant effects
Catalog No. 50-004-32918 Supplier Medchemexpress LLC Supplier No. HYB0114R100MG
Oxcarbazepine (Standard) is the analytical standard of Oxcarbazepine, intended for research and analytical applications. It is a sodium channel blocker that significantly inhibits glioblastoma cell growth and induces apoptosis or G2/M arrest in glioblastoma cell lines. It exhibits anti-cancer and anticonvulsant effects. This product is for research use only.
  • Analytical standard
  • Used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS
  • Sodium channel blocker
  • Inhibits glioblastoma cell growth
  • Induces apoptosis or G2/M arrest in glioblastoma cell lines
  • Anti-cancer effects
  • Anticonvulsant effects