Learn More
Medchemexpress LLC Oxcarbazepine (Standard) | 28721-07-5 | 99.9% | C₁₅H₁₂N₂O₂ | 25 MG
SDP
Catalog No. 5000358732
Encompass_Preferred
Oxcarbazepine (Standard) is an analytical standard intended for research and analytical applications. It functions as a sodium channel blocker, significantly inhibiting glioblastoma cell growth and inducing apoptosis or G2/M arrest in glioblastoma cell lines. It exhibits both anti-cancer and anticonvulsant effects.
  • Intended for research and analytical applications.
  • Functions as a sodium channel blocker.
  • Significantly inhibits glioblastoma cell growth.
  • Induces apoptosis or G2/M arrest in glioblastoma cell lines.
  • Exhibits anti-cancer and anticonvulsant effects.
  • Used as a reference standard for assays.
  • Commonly used in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.
Catalog No. 50-003-58732 Supplier Medchemexpress LLC Supplier No. 25MGHYB0114R
Oxcarbazepine (Standard) is an analytical standard intended for research and analytical applications. It functions as a sodium channel blocker, significantly inhibiting glioblastoma cell growth and inducing apoptosis or G2/M arrest in glioblastoma cell lines. It exhibits both anti-cancer and anticonvulsant effects.
  • Intended for research and analytical applications.
  • Functions as a sodium channel blocker.
  • Significantly inhibits glioblastoma cell growth.
  • Induces apoptosis or G2/M arrest in glioblastoma cell lines.
  • Exhibits anti-cancer and anticonvulsant effects.
  • Used as a reference standard for assays.
  • Commonly used in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.