Learn More
Medchemexpress LLC Oxcarbazepine (Standard) | 28721-07-5 | 99.9% | C₁₅H₁₂N₂O₂ | 5 MG
SDP
Catalog No. 5000432428
Encompass_Preferred
Oxcarbazepine (Standard) is the analytical standard of Oxcarbazepine, intended for research and analytical applications. It acts as a sodium channel blocker and significantly inhibits glioblastoma cell growth, inducing apoptosis or G2/M arrest in glioblastoma cell lines. It exhibits anti-cancer and anticonvulsant effects.
  • Analytical standard grade
  • Acts as a sodium channel blocker
  • Significantly inhibits glioblastoma cell growth
  • Induces apoptosis or G2/M arrest in glioblastoma cell lines
  • Exhibits anti-cancer effects
  • Exhibits anticonvulsant effects
  • Commonly used as a reference standard for assays in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.
Catalog No. 50-004-32428 Supplier Medchemexpress LLC Supplier No. HYB0114R5MG
Add to Cart
Add to Cart
Oxcarbazepine (Standard) is the analytical standard of Oxcarbazepine, intended for research and analytical applications. It acts as a sodium channel blocker and significantly inhibits glioblastoma cell growth, inducing apoptosis or G2/M arrest in glioblastoma cell lines. It exhibits anti-cancer and anticonvulsant effects.
  • Analytical standard grade
  • Acts as a sodium channel blocker
  • Significantly inhibits glioblastoma cell growth
  • Induces apoptosis or G2/M arrest in glioblastoma cell lines
  • Exhibits anti-cancer effects
  • Exhibits anticonvulsant effects
  • Commonly used as a reference standard for assays in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.