Learn More
Aviva Systems Biology Corp OXCT1 Antibody - middle region (ARP48481_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48481_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against OXCT1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human OXCT1. Alternate names: OXCT, SCOTApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-53556