Learn More
Matrix Scientific Oxetane-3-carboxylic acid, 114012-41-8, MFCD08669505, 250mg
Supplier: Matrix Scientific 066986250MG
Molecular Formula C4H6O3, Molecular Weight 102.09, Boiling Point 244.2
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.