Learn More
Medchemexpress LLC Oxetane, 3-iodo- | 26272-85-5 | MFCD08544403 | >95.0% | 183.98 g/mol | C3H5IO | 10 G
SDP
Catalog No. 5000302158
Encompass_Preferred
3-Iodooxetane (CAS 26272-85-5) is an organoiodide oxetane used as a synthetic building block and biochemical reagent in organic and medicinal chemistry. It is typically supplied as a high-purity, colorless liquid for small-scale laboratory research and is used to introduce an iodo substituent for further functionalization.
  • High-purity material suitable for research use.
  • Useful building block for organic synthesis and medicinal chemistry.
  • Facilitates nucleophilic substitution and ring-opening reactions.
  • Colorless liquid with formula C3H5IO and molecular weight 183.98 g/mol.
  • Small-scale packaging convenient for laboratory workflows.
  • Store cold and protect from light to preserve stability.
Catalog No. 50-003-02158 Supplier Medchemexpress LLC Supplier No. HYW01630610G
3-Iodooxetane (CAS 26272-85-5) is an organoiodide oxetane used as a synthetic building block and biochemical reagent in organic and medicinal chemistry. It is typically supplied as a high-purity, colorless liquid for small-scale laboratory research and is used to introduce an iodo substituent for further functionalization.
  • High-purity material suitable for research use.
  • Useful building block for organic synthesis and medicinal chemistry.
  • Facilitates nucleophilic substitution and ring-opening reactions.
  • Colorless liquid with formula C3H5IO and molecular weight 183.98 g/mol.
  • Small-scale packaging convenient for laboratory workflows.
  • Store cold and protect from light to preserve stability.