Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Oxetane-3-methanol

Catalog No. 745715250MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
74-571-5250MG 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 74-571-5250MG Supplier Sigma Aldrich Supplier No. 745715250MG
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Oxetane-3-methanol

Specifications

Density 1.092 g/mL (at 25°C)
Molecular Formula C4H8O2
Refractive Index n20/D 1.450
Linear Formula C4H8O2
MDL Number MFCD17015894
Quantity 100 g
Synonym 3-(Hydroxymethyl)oxetane; 3-Oxetanemethanol; 3-Oxetanylmethanol
Molecular Weight (g/mol) 88.11
Percent Purity 97%
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.