Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oxethazaine, MilliporeSigma™ Supelco™

Catalog No. 111012854 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2854 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-2854 Supplier MilliporeSigma Supelco Supplier No. O53805G
Add to Cart
Add to Cart

Oxethazaine is a topical anesthetic, administered as an antacid for treating pain associated with duodenal ulcer, esophagitis and gastritis.

TRUSTED_SUSTAINABILITY

Specifications

CAS 126-27-2
Physical Form Neat
Linear Formula C28H41N3O3
MDL Number MFCD00079202
Synonym 2-Hydroxyethyliminobis(N-[alpha,alpha-dimethylphenethyl]-N-methylacetamide); Oxetacaine
Type Analytical Standard
Quantity 5 g
Formula Weight 467.64
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.