Learn More
TARGETMOL CHEMICALS INC OXF BD 02 1MG
SDP
Catalog No. 502876278
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. OXF BD 02 is a potent and selective BRD4(1) inhibitor (IC50 382 nM) with anticancer and anti-inflammatory activity. Purity 98.25%
Catalog No. 50-287-6278 Supplier TARGETMOL CHEMICALS INC Supplier No. T231151MG
Add to Cart
Add to Cart
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. OXF BD 02 is a potent and selective BRD4(1) inhibitor (IC50 382 nM) with anticancer and anti-inflammatory activity. Purity 98.25%