Learn More
Medchemexpress LLC Oxfbd04 | 2231747-03-6 | 99.0% | C17H16N2O3 | 10MG
SDP
Catalog No. 5000258607
Encompass_Preferred
OXFBD04 is a small-molecule BET bromodomain ligand that inhibits BRD4 and shows modest affinity for the CREBBP bromodomain. It is intended for research use and has reported analytical data including CAS 2231747-03-6, molecular formula C17H16N2O3, molecular weight 296.32, and reported purity ~98.99%.
  • Potent BRD4 inhibitor for epigenetic research.
  • Modest affinity for the CREBBP bromodomain.
  • High reported purity (≈98.99%).
  • Soluble ≥250 mg/mL in DMSO for formulation convenience.
  • Available in small pack sizes suitable for screening.
  • Storage conditions recommended for powder and solution stability.
Catalog No. 50-002-58607 Supplier Medchemexpress LLC Supplier No. HY13523610MG
OXFBD04 is a small-molecule BET bromodomain ligand that inhibits BRD4 and shows modest affinity for the CREBBP bromodomain. It is intended for research use and has reported analytical data including CAS 2231747-03-6, molecular formula C17H16N2O3, molecular weight 296.32, and reported purity ~98.99%.
  • Potent BRD4 inhibitor for epigenetic research.
  • Modest affinity for the CREBBP bromodomain.
  • High reported purity (≈98.99%).
  • Soluble ≥250 mg/mL in DMSO for formulation convenience.
  • Available in small pack sizes suitable for screening.
  • Storage conditions recommended for powder and solution stability.