Learn More
TARGETMOL CHEMICALS INC OXFBD04 5MG
SDP
Catalog No. 502581464
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. OXFBD04 is a potent and selective BRD4 inhibitor (IC50= 166 nM). OXFBD04 is a potent BET bromodomain ligand and exhibits additional modest affinity for the CREBBP bromodomain. OXFBD04 exhibits anti-cancer activity. purity: 98%
Catalog No. 50-258-1464 Supplier TARGETMOL CHEMICALS INC Supplier No. T123385MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. OXFBD04 is a potent and selective BRD4 inhibitor (IC50= 166 nM). OXFBD04 is a potent BET bromodomain ligand and exhibits additional modest affinity for the CREBBP bromodomain. OXFBD04 exhibits anti-cancer activity. purity: 98%