Learn More
Cayman Chemical Oxfendazole, Purity: ≥98%, CAS: 53716-50-0, 5 mg
SDP
Catalog No. 502989038
Encompass_Preferred
An anthelmintic; curative in pig models of H. rubidus or A. suum infection at 3 and 6 mg/kg, respectively; is lethal to H. contortus and T. circumcincta when administered to infected cattle (LD50s = 4.28 and 3.61 mg/kg of host weight, respectively)
Catalog No. 50-298-9038 Supplier Cayman Chemical Supplier No. 297425 MG
An anthelmintic; curative in pig models of H. rubidus or A. suum infection at 3 and 6 mg/kg, respectively; is lethal to H. contortus and T. circumcincta when administered to infected cattle (LD50s = 4.28 and 3.61 mg/kg of host weight, respectively)