Learn More
JDM LAB SOLUTIONS LLC Oxford BenchMate Microplate Shaker
Catalog No. NC2385865
Encompass
2mm Orbital Diameter with a powerful motor that allows up to 1800 RPM!Brushless DC motor for quiet operation and long-life operation with no maintenance.Microprocessor-controlled unit with digital display.Every unit comes with replaceable clamps that allow for compatibility with a variety of standard microplates.Every unit also includes a microtube attachment for 1.5mL or 2.0mL Microtubes.
Catalog No. NC2385865 Supplier JDM LAB SOLUTIONS LLC Supplier No. SPM
2mm Orbital Diameter with a powerful motor that allows up to 1800 RPM!Brushless DC motor for quiet operation and long-life operation with no maintenance.Microprocessor-controlled unit with digital display.Every unit comes with replaceable clamps that allow for compatibility with a variety of standard microplates.Every unit also includes a microtube attachment for 1.5mL or 2.0mL Microtubes.