Learn More
JDM LAB SOLUTIONS LLC Oxford Benchmate Digital Tube Rotator
Catalog No. NC2385826
Encompass
3 plate options available: Supplied with any one disc plate of your choice.Adjustable speed ranging from 10-80 rpm in steps of, and with an accuracy of, +/- 1 rpm.Timer setting from 1 to 99 minutes, and infinite mode.The angle can be adjusted from 0 to 90 degrees.Programmable – The user can set and save up to 9 user-defined programs (protocols).Option for auto power on in case of power failure – on the resumption of power the unit will start automatically and run for the remaining runtime.Easy removal and installation of disc plates for replacement and cleaning.
Catalog No. NC2385826 Supplier JDM LAB SOLUTIONS LLC Supplier No. OTR24D
3 plate options available: Supplied with any one disc plate of your choice.Adjustable speed ranging from 10-80 rpm in steps of, and with an accuracy of, +/- 1 rpm.Timer setting from 1 to 99 minutes, and infinite mode.The angle can be adjusted from 0 to 90 degrees.Programmable – The user can set and save up to 9 user-defined programs (protocols).Option for auto power on in case of power failure – on the resumption of power the unit will start automatically and run for the remaining runtime.Easy removal and installation of disc plates for replacement and cleaning.