Learn More
JDM LAB SOLUTIONS LLC OXFORD C8-M MICRO CENTRIFUGE
Catalog No. NC2127782
Encompass
The C8 Microcentrifuge has a fixed speed of 6,000 rpm / 2,000 x g and a noise level of < 55 dB. It is ideal for all quick spin centrifugation applications.
Catalog No. NC2127782 Supplier JDM LAB SOLUTIONS LLC Supplier No. C8M
The C8 Microcentrifuge has a fixed speed of 6,000 rpm / 2,000 x g and a noise level of < 55 dB. It is ideal for all quick spin centrifugation applications.