Learn More
Bioworld Oxford Listeria Agar, 500 g
Supplier:  Bioworld 306200571
Encompass_Preferred
Oxford Listeria AgarUsed with addition of antibiotics for the selective isolation of Listeria monocytogenes from foodpH: 7.2Working Concentration: 55.5 g/LDehydrated Appearance: beige, homogeneous powderPrepared Appearance: light to medium amber agarSoluble in boiling waterContains:Columbia Blood Agar Base Esculin Ferric Ammonium Citrate Lithium Chloride AgarPer L of media, disolve the following in 10 mL of a 50% solution of ethanol.Cycloheximide - 0.4 gColistin sulfide - 0.02 gFosfomycin - 0.01 gAcriflavine - 5.0 mgCerofotetan - 2.0 mgFilter sterilize antibiotic mix and add to L of autoclaved media after it has cooled.
Catalog No. 50-254-343