Learn More
Oxyrase Oxyrase For Broth 10 Ml Dropper Bottle, 6/Pk
SDP
Catalog No. 502005298
Encompass
Oxyrase for Broth produces anaerobic conditions in wide variety of bacteriological broth media. Makes growing anaerobes easy; especially reviving lyophiles. Add Oxyrase for Broth to your broth medium. Broth goes anaerobic in 10-15 minutes with continual oxygen removal. Never boil broth tubes again.
Catalog No. 50-200-5298 Supplier Oxyrase Supplier No. OB0010
Oxyrase for Broth produces anaerobic conditions in wide variety of bacteriological broth media. Makes growing anaerobes easy; especially reviving lyophiles. Add Oxyrase for Broth to your broth medium. Broth goes anaerobic in 10-15 minutes with continual oxygen removal. Never boil broth tubes again.