Learn More
Enzo Life Sciences OXI-TEK TBARS assay kit (160 tests)
SDP
Supplier:  Enzo Life Sciences ALX850287KI01
Encompass_Preferred
The sensitivity of measuring thiobarbituric acid reactive substances (TBARS) has made this assay the method of choice for screening and monitoring lipid peroxidation, which is a major indication of oxidative stress. Malondialdehyde (MDA) forms a 1:2 adduct with thiobarbituric acid; the adduct can be measured by fluorometry. Biological specimens contain a mixture of TBARS, including lipid hydroperoxides and aldehydes. In practice, TBARS are expressed in equivalent numbers to MDA. Applications: Fluorescence microscopy .
Catalog No. 50-201-0152