Learn More
Cayman Chemical OxIbendazole 100mg
SDP
Catalog No. 502940723
Encompass_Preferred
A benzimidazole anthelmintic; inhibits binding of [3H]mebendazole to H. contortus L3 larval tubulin and inhibits mammalian tubulin polymerization (IC50s = 0.3 and 2.2 µM, respectively); reduces fecal worm egg counts in dogs and cats by greater than 90% for the nematodes T. canis, T. vulpis, A. caninum, and T. leonine at 15 mg/kg in combination with niclosamide; reduces fecal egg counts of strongyles in horses at 10 mg/kg per day
Catalog No. 50-294-0723 Supplier Cayman Chemical Supplier No. 27365100 MG
A benzimidazole anthelmintic; inhibits binding of [3H]mebendazole to H. contortus L3 larval tubulin and inhibits mammalian tubulin polymerization (IC50s = 0.3 and 2.2 µM, respectively); reduces fecal worm egg counts in dogs and cats by greater than 90% for the nematodes T. canis, T. vulpis, A. caninum, and T. leonine at 15 mg/kg in combination with niclosamide; reduces fecal egg counts of strongyles in horses at 10 mg/kg per day