Learn More
Medchemexpress LLC Oxibendazole | 20559-55-1 | 99.6% | 25 MG
SDP
Catalog No. 5000450553
Encompass_Preferred
Oxibendazole is an effective benzimidazole anthelmintic that combats nematode infections. It has demonstrated the ability to induce apoptosis and exhibits both anti-cancer and anti-inflammation activities.
  • Induces apoptosis
  • Active against nematode infections
  • Exhibits anti-cancer activities
  • Possesses anti-inflammation properties
  • Downregulates phosphorylation of ERK1/2, P90RSK, and S6
  • Upregulates expression of phosphorylated JNK, AKT, and P70S6K
Catalog No. 50-004-50553 Supplier Medchemexpress LLC Supplier No. 25MGHYB0299
Oxibendazole is an effective benzimidazole anthelmintic that combats nematode infections. It has demonstrated the ability to induce apoptosis and exhibits both anti-cancer and anti-inflammation activities.
  • Induces apoptosis
  • Active against nematode infections
  • Exhibits anti-cancer activities
  • Possesses anti-inflammation properties
  • Downregulates phosphorylation of ERK1/2, P90RSK, and S6
  • Upregulates expression of phosphorylated JNK, AKT, and P70S6K