Learn More
TARGETMOL CHEMICALS INC Oxiconazole nitrate 25MG
SDP
Catalog No. 502878357
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 50 mg, 100 mg, 200 mg, 500 mg and bulk. Please contact Fisher for quotes. Oxiconazole nitrate (Ro 13-8996) is the nitrate salt form of oxiconazole, a broad-spectrum imidazole derivative with antifungal activity. Likely inhibiting cytochrome P450-dependent demethylation of lanosterol, it prevents ergosterol synthesis, a crucial component of the fungal cell membrane. This disruption of membrane synthesis and integrity alters permeability, promotes loss of essential intracellular components, and inhibits fungal cell growth. Purity 99.73%
Catalog No. 50-287-8357 Supplier TARGETMOL CHEMICALS INC Supplier No. T129025MG
Also available in 1 mL, 5 mg, 10 mg, 50 mg, 100 mg, 200 mg, 500 mg and bulk. Please contact Fisher for quotes. Oxiconazole nitrate (Ro 13-8996) is the nitrate salt form of oxiconazole, a broad-spectrum imidazole derivative with antifungal activity. Likely inhibiting cytochrome P450-dependent demethylation of lanosterol, it prevents ergosterol synthesis, a crucial component of the fungal cell membrane. This disruption of membrane synthesis and integrity alters permeability, promotes loss of essential intracellular components, and inhibits fungal cell growth. Purity 99.73%