Learn More
Apexbio Technology LLC Oxiconazole (nitrate) 64211-46-7 50mg
SDP
Catalog No. 5000089490
Encompass_Preferred
Oxiconazole nitrate (CAS 64211-46-7) is an imidazole derivative with antifungal properties It is designed to inhibit ergosterol synthesis thereby perturbing fungal cell membrane integrity and cellular functions Oxiconazole nitrate exerts its biological activity primarily through inhibition of ergosterol synthesis in fungal cells In in vitro assays oxiconazole nitrate demonstrates fungistatic activity against human pathogenic dermatophytes including Trichophyton rubrum T mentagrophytes and Epidermophyton floccosum with IC50 values typically in the submicromolar range Based on these pharmacological properties oxiconazole nitrate holds research potential in the treatment of fungal infections such as trichophytosis and vaginal candidiasis and has been evaluated in both topical and systemic experimental models
Catalog No. 50-000-89490 Supplier Apexbio Technology LLC Supplier No. C341850
Oxiconazole nitrate (CAS 64211-46-7) is an imidazole derivative with antifungal properties It is designed to inhibit ergosterol synthesis thereby perturbing fungal cell membrane integrity and cellular functions Oxiconazole nitrate exerts its biological activity primarily through inhibition of ergosterol synthesis in fungal cells In in vitro assays oxiconazole nitrate demonstrates fungistatic activity against human pathogenic dermatophytes including Trichophyton rubrum T mentagrophytes and Epidermophyton floccosum with IC50 values typically in the submicromolar range Based on these pharmacological properties oxiconazole nitrate holds research potential in the treatment of fungal infections such as trichophytosis and vaginal candidiasis and has been evaluated in both topical and systemic experimental models