Learn More
Medchemexpress LLC Oxiconazole nitrate | 64211-46-7 | 98.0% | 100 MG
SDP
Catalog No. 5000412079
Encompass_Preferred
Oxiconazole nitrate is a broad-spectrum anti-fungal agent effective against Candida, Aspergillus, and Trichophyton. It acts as a highly efficacious activator of CYP3A4 transactivation and demonstrates an inhibitory effect against colorectal cancer (CRC) by peroxiredoxin-2 (PRDX2)-mediated autophagy arrest. This product is intended for research use only.
  • Broad-spectrum anti-fungal agent against Candida, Aspergillus, and Trichophyton.
  • Activator of CYP3A4 transactivation.
  • Exhibits inhibitory effect against colorectal cancer (CRC).
  • Inhibits CRC cell growth and shows no obvious side effects in animal models.
Catalog No. 50-004-12079 Supplier Medchemexpress LLC Supplier No. HYB1324100MG
Oxiconazole nitrate is a broad-spectrum anti-fungal agent effective against Candida, Aspergillus, and Trichophyton. It acts as a highly efficacious activator of CYP3A4 transactivation and demonstrates an inhibitory effect against colorectal cancer (CRC) by peroxiredoxin-2 (PRDX2)-mediated autophagy arrest. This product is intended for research use only.
  • Broad-spectrum anti-fungal agent against Candida, Aspergillus, and Trichophyton.
  • Activator of CYP3A4 transactivation.
  • Exhibits inhibitory effect against colorectal cancer (CRC).
  • Inhibits CRC cell growth and shows no obvious side effects in animal models.