Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Oxiconazole nitrate | 64211-46-7 | MFCD01727545 | 98.0% | 492.14 | C18H14Cl4N4O4 | 10 MG
SDP

Catalog No. 5000297206
Encompass_Preferred

Oxiconazole nitrate is an imidazole-class antifungal research compound supplied as a high-purity solid for laboratory use. The nitrate salt of oxiconazole shows broad-spectrum activity against dermatophytes and common fungal pathogens and is used in biochemical screening and in vitro antifungal assays.

  • Chemical formula C18H14Cl4N4O4.
  • CAS number 64211-46-7.
  • Molecular weight 492.14 g/mol.
  • Reported purity 98.0%.
  • Suitable for in vitro antifungal studies and screening assays.
  • Available as a 10 mg solid for small-scale research use.

Catalog No. 50-002-97206 Supplier Medchemexpress LLC Supplier No. HYB132410MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Oxiconazole nitrate is an imidazole-class antifungal research compound supplied as a high-purity solid for laboratory use. The nitrate salt of oxiconazole shows broad-spectrum activity against dermatophytes and common fungal pathogens and is used in biochemical screening and in vitro antifungal assays.

  • Chemical formula C18H14Cl4N4O4.
  • CAS number 64211-46-7.
  • Molecular weight 492.14 g/mol.
  • Reported purity 98.0%.
  • Suitable for in vitro antifungal studies and screening assays.
  • Available as a 10 mg solid for small-scale research use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.