Learn More
VEOLIA WTS Oxidizer Cartridge, Low-capacity
Catalog No. NC0703915
Encompass
Low-capacity, 150-mL, Oxidizer Reagent Cartridge for the Sievers M-Series. Replace after 3 months of use.
Catalog No. NC0703915 Supplier VEOLIA WTS Supplier No. APF 9015002
Low-capacity, 150-mL, Oxidizer Reagent Cartridge for the Sievers M-Series. Replace after 3 months of use.