Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oxidation Reagent, Honeywell Burdick & Jackson™

Catalog No. p-7145560 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
4 x 2 L
4 x 2.5 L
56 L
200 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-090-428 4 x 2 L
60-090-001 4 x 2.5 L
60-090-432 56 L
60-090-431 200 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 60-090-428 Supplier Honeywell Chemicals Supplier No. BR6662
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

BioSyn™, suitable for use with DNA/RNA synthesis instruments, 0.02M Iodine in 70/20/10 THF/Pyridine/Water (v/v)

Burdick & Jackson® BioSyn® reagents & solvents for DNA/RNA synthesis

  • Honeywell Burdick & Jackson® BioSyn® Solvents and Reagents are specifically processed and purified to exacting specifications, ensuring outstanding synthesis efficiency
  • ideal for oligonucleotide synthesis regardless of the application
  • All solvents and reagents are filtered (0.2-micron for oxidation reagents and 0.1-micron for all other reagents) to minimize particulate contamination

 


 

TRUSTED_SUSTAINABILITY

Specifications

CAS 7553-56-2, 109-99-9, 7732-18-5, 110-86-1
CAS Max % 0.50%, 67.20%, 10.90%, 21.40%
Quantity 4 x 2 L
Flash Point -15°C
Packaging Glass Bottle
Physical Form Liquid, clear
Purity Grade Notes Oxidation reagents
Boiling Point 66°C
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.