Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oxidation Reagent, Solstice

Catalog No. 60048527 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
4 L
10 L
20 L
200 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-048-527 10 L
60-048-524 4 L
60-090-427 20 L
60-090-426 200 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 60-048-527 Supplier Solstice Supplier No. SR66610
This item is not returnable. View return policy

BioSyn™, suitable for use with DNA/RNA synthesis instruments, 0.02M Iodine in 70/20/10 THF/Pyridine/Water (v/v)

TRUSTED_SUSTAINABILITY

Specifications

CAS 109-99-9, 110-86-1, 7732-18-5, 7553-56-2
CAS Max % 0.50%, 67.20%, 10.90%, 21.40%
Quantity 10 L
Flash Point -15°C
Packaging Returnable Stainless Steel Drum
Physical Form Liquid, clear
Purity Grade Notes Oxidation reagents
Boiling Point 66°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.