Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oxidation Reagent,Solstice

Catalog No. 60090004 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
4X2.5L GLASS BOTTLE
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
60-090-004 4X2.5L GLASS BOTTLE
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-090-004 Supplier Solstice Supplier No. BR6634X2.5L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BioSyn™, suitable for use with DNA/RNA synthesis instruments, 0.02M Iodine in 78/20/2 THF/Pyridine/Water (v/v)

Solstice BioSyn™ reagents and solvents for DNA/RNA synthesis

  • Solstice BioSyn™ Solvents and Reagents are specifically processed and purified to exacting specifications, ensuring outstanding synthesis efficiency
  • ideal for oligonucleotide synthesis regardless of the application
  • All solvents and reagents are filtered (0.2-micron for oxidation reagents and 0.1-micron for all other reagents) to minimize particulate contamination

TRUSTED_SUSTAINABILITY

Specifications

CAS 109-99-9, 110-86-1, 7732-18-5, 7553-56-2
CAS Max % 78.00%, 20.00%, 2.00%, 0.20%
Quantity 4 x 2.5 L
Flash Point -15°C
Packaging Glass Bottle
Physical Form Liquid, clear
Purity Grade Notes Oxidation reagents
Boiling Point 66°C
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.