Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oxidation Reagent, Solstice

Catalog No. 60090005 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
20 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-090-005 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-090-005 Supplier Solstice Supplier No. BN673AK20
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BioSyn™, suitable for use with DNA/RNA synthesis instruments, 0.025M Iodine in 45/45/10 Pyridine/Acetonitrile/Water (v/v/v)

TRUSTED_SUSTAINABILITY

Specifications

CAS 7732-18-5, 7553-56-2, 110-86-1, 75-05-8
CAS Max % 11.10%, 0.60%, 49.10%, 39.20%
Quantity 20 L
Flash Point 8.89°C
Packaging Returnable NOWPAK™ II
Physical Form Liquid
Purity Grade Notes Oxidation reagents
Boiling Point 80.56°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.