Learn More
Cayman Chemical OxIdatIve Stres MaxSpec LC 1ea
SDP
Catalog No. 502966158
Encompass_Preferred
A mixture of lipids and nucleic acids produced during oxidative stress in biological systems; designed for direct use in LC-MS applications
Catalog No. 50-296-6158 Supplier Cayman Chemical Supplier No. 187011 EA
A mixture of lipids and nucleic acids produced during oxidative stress in biological systems; designed for direct use in LC-MS applications