Learn More
Electron Microscopy Sciences Oxide Remover Cleaner, 1 Gallon 1 GL
SDP
Catalog No. 50930736
Encompass_Preferred
This cleaner may be used on brass, copper, steel and other metals to remove rust and metal-oxides. Comes as a concentrate. Dilute to 10 – 12% with water for use with ultrasonic machine.
Catalog No. 50-930-736 Supplier Electron Microscopy Sciences Supplier No. 71053
This cleaner may be used on brass, copper, steel and other metals to remove rust and metal-oxides. Comes as a concentrate. Dilute to 10 – 12% with water for use with ultrasonic machine.