Learn More
HCL Labels Oxidizers - Cabinet or Secondary Containment Sign - 3.5" x 11" Adhesive Vinyl (Pack of 5)
SDP
Catalog No. NC1977731
Encompass_Preferred
Oxidizers Cabinet or Secondary Containment Label with GHS pictogram - Pack of 5 LabelsLABELS FOR CHEMICAL STORAGE CABINETS OR SECONDARY CONTAINER TRAYS•With GHS pictograms for quick hazard identification•Assists scientist with the proper segregation of chemicals•Printed using chemical resistant adhesive vinyl•Air egress adhesive that makes for an easy bubble-free application
Catalog No. NC1977731 Supplier HCL Labels Supplier No. SSLX216VN35115
Oxidizers Cabinet or Secondary Containment Label with GHS pictogram - Pack of 5 LabelsLABELS FOR CHEMICAL STORAGE CABINETS OR SECONDARY CONTAINER TRAYS•With GHS pictograms for quick hazard identification•Assists scientist with the proper segregation of chemicals•Printed using chemical resistant adhesive vinyl•Air egress adhesive that makes for an easy bubble-free application