Learn More
Medchemexpress LLC Oxidized low density lipoprotein (mouse) | ≥98% | 1 MG
SDP
Catalog No. 5000426629
Encompass_Preferred
Oxidized low density lipoprotein (mouse) (Mouse ox-LDL) is an oxidized low density lipoprotein (LDL). It induces atherosclerosis (AS) by facilitating endothelial dysfunction and accelerating the VSMCs growth and migration, making it suitable for constructing an in vitro model of AS.
  • Induces atherosclerosis (AS)
  • Facilitates endothelial dysfunction
  • Accelerates VSMCs growth and migration
  • Can be used to construct an in vitro model of AS
  • Upregulates mRNA and protein expressions of long noncoding RNA TUG1 and receptor tyrosine kinase-like orphan receptor 2 (ROR2)
  • Promotes the proliferation of HA-VSMCs
  • Enhances migration and invasion abilities
  • Increases the expressions of metastasis-related proteins β-catenin and Vimentin in human aortic vascular smooth muscle cells
Catalog No. 50-004-26629 Supplier Medchemexpress LLC Supplier No. HYNP0131MG
Oxidized low density lipoprotein (mouse) (Mouse ox-LDL) is an oxidized low density lipoprotein (LDL). It induces atherosclerosis (AS) by facilitating endothelial dysfunction and accelerating the VSMCs growth and migration, making it suitable for constructing an in vitro model of AS.
  • Induces atherosclerosis (AS)
  • Facilitates endothelial dysfunction
  • Accelerates VSMCs growth and migration
  • Can be used to construct an in vitro model of AS
  • Upregulates mRNA and protein expressions of long noncoding RNA TUG1 and receptor tyrosine kinase-like orphan receptor 2 (ROR2)
  • Promotes the proliferation of HA-VSMCs
  • Enhances migration and invasion abilities
  • Increases the expressions of metastasis-related proteins β-catenin and Vimentin in human aortic vascular smooth muscle cells