Learn More
Sigma Aldrich Fine Chemicals Biosciences Iodine Solution | 7553-56-2 | 4 L | Oxidizing Reagent
Catalog No. NC2611338
Encompass
Iodine solution, 0.05M in pyridine | Purity: 0.05M | Mol Wt: 253.81 | CAS No: 7553-56-2 | Oxidizing reagent | 4L
Catalog No. NC2611338 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. BI0424P4000
Iodine solution, 0.05M in pyridine | Purity: 0.05M | Mol Wt: 253.81 | CAS No: 7553-56-2 | Oxidizing reagent | 4L