Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Oxidizing Reagent(0.5M), I2 in Pyridine

Catalog No. MBI0424NP20 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
20 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MBI0424NP20 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MBI0424NP20 Supplier MilliporeSigma™ Supplier No. BI0424NP20
Add to Cart
Add to Cart
Request a Quote

Used for synthesis of DNA

  • Accessory for biosynthesis of DNA
TRUSTED_SUSTAINABILITY

Specifications

Color Dark brown
Concentration 89 to 91.0%
Formula Weight Mixture
Packaging Type Plastic barrel
Product Type Oxidizing reagent
Quantity 20 L
Synonym Oxidizing Reagent
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.