Learn More
Sigma Aldrich Fine Chemicals Biosciences Iodine monoxide | 12029-98-0 | MFCD00003543 | 4 L | 0.02 M in Tetrahydrofuran/Pyridine/Water (70:20:10)
Catalog No. NC2468997
Encompass
Iodine monoxide | Purity: Not specified | Mol Wt: 227.89 | CAS No: 12029-98-0 | MDL No: MFCD00003543 | 4LThis iodine monoxide solution can function as an oxidizing agent for oxidation reactions in organic synthesis. The 0.02 molar I2O is dissolved in a 70:20:10 volume ratio mixture of tetrahydrofuran, pyridine, and water solvents. The product is provided in 4 liter poly coated glass bottles.
Catalog No. NC2468997 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. BI0420P4000
Iodine monoxide | Purity: Not specified | Mol Wt: 227.89 | CAS No: 12029-98-0 | MDL No: MFCD00003543 | 4LThis iodine monoxide solution can function as an oxidizing agent for oxidation reactions in organic synthesis. The 0.02 molar I2O is dissolved in a 70:20:10 volume ratio mixture of tetrahydrofuran, pyridine, and water solvents. The product is provided in 4 liter poly coated glass bottles.