Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oxidizing Reagent, 02M I2O in THF/Pyridine/Water, Novabiochem™, MilliporeSigma™

Catalog No. MBI04204000 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
4 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MBI04204000 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MBI04204000 Supplier MilliporeSigma Supplier No. BI04204000
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recommended Storage

Store below +30°C.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula Mixture
Synonym DNA Reagent

Specifications

Color Red
Packaging Glass Bottle
Quantity 4 L
Physical Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.