Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

[(Oxido)phenyl(trifluoromethyl)-lambda4-sulfanylidene]dimethylammonium Tetrafluoroborate 98.0+%, TCI America™
SDP

Catalog No. O03671G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O03671G 1 g
O0367200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O03671G Supplier TCI America Supplier No. O03671G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1046786-08-6
Molecular Formula C9H11BF7NOS
Synonym Shibata Reagent I

Specifications

Color White
Quantity 1 g
UN Number 1759
Formula Weight 325.05
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material [(Oxido)phenyl(trifluoromethyl)-lambda4-sulfanylidene]dimethylammonium Tetrafluoroborate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.