Learn More
Selleck Chemical LLC Oxidopamine (hydrobromide) 25mg 636-00-0 6-hydroxydopamine; 6-OHDA; 2,4,5-trihydroxyphenethylamine
SDP
Supplier:  Selleck Chemical LLC S532425MG
Encompass_Preferred
Oxidopamine (6-hydroxydopamine, 6-OHDA, 2,4,5-trihydroxyphenethylamine) is a neurotoxic synthetic organic compound that acts as an antagonist of the neurotransmitter dopamine with potential antineoplastic activity. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8364