Learn More
Medchemexpress LLC Oxidopamine (hydrobromide) | 636-00-0 | 99.4% | 500 MG
SDP
Catalog No. 5000413860
Encompass_Preferred
Oxidopamine (6-OHDA) hydrobromide is a potent neurotoxin that selectively destroys dopaminergic neurons by acting as an antagonist of the neurotransmitter dopamine. It is a valuable tool for research into various neurological conditions, including Parkinson's disease (PD), attention-deficit hyperactivity disorder (ADHD), and Lesch-Nyhan syndrome. The compound also promotes COX-2 activation, leading to PGE2 synthesis and pro-inflammatory cytokine IL-1β secretion.
  • Selectively destroys dopaminergic neurons
  • Antagonist of dopamine neurotransmitter
  • Promotes COX-2 activation and PGE2 synthesis
  • Induces pro-inflammatory cytokine IL-1β secretion
  • Useful for Parkinson's disease research models
  • Applicable in ADHD and Lesch-Nyhan syndrome studies
Catalog No. 50-004-13860 Supplier Medchemexpress LLC Supplier No. HYB1081A500MG
Oxidopamine (6-OHDA) hydrobromide is a potent neurotoxin that selectively destroys dopaminergic neurons by acting as an antagonist of the neurotransmitter dopamine. It is a valuable tool for research into various neurological conditions, including Parkinson's disease (PD), attention-deficit hyperactivity disorder (ADHD), and Lesch-Nyhan syndrome. The compound also promotes COX-2 activation, leading to PGE2 synthesis and pro-inflammatory cytokine IL-1β secretion.
  • Selectively destroys dopaminergic neurons
  • Antagonist of dopamine neurotransmitter
  • Promotes COX-2 activation and PGE2 synthesis
  • Induces pro-inflammatory cytokine IL-1β secretion
  • Useful for Parkinson's disease research models
  • Applicable in ADHD and Lesch-Nyhan syndrome studies