Learn More
Medchemexpress LLC Oxidopamine hydrobromide | 636-00-0 | 99.42% | 100 MG
SDP
Catalog No. 5000413976
Encompass_Preferred
Oxidopamine (6-OHDA) hydrobromide is an antagonist of the neurotransmitter dopamine. It is a widely used neurotoxin that selectively destroys dopaminergic neurons. It promotes COX-2 activation, leading to PGE2 synthesis and pro-inflammatory cytokine IL-1β secretion. Oxidopamine hydrobromide can be used for the research of Parkinson's disease (PD), attention-deficit hyperactivity disorder (ADHD), and Lesch-Nyhan syndrome.
  • Antagonist of the neurotransmitter dopamine.
  • Widely used neurotoxin.
  • Selectively destroys dopaminergic neurons.
  • Promotes COX-2 activation.
  • Leads to PGE2 synthesis and pro-inflammatory cytokine IL-1β secretion.
  • Used for research of Parkinson's disease (PD), attention-deficit hyperactivity disorder (ADHD), and Lesch-Nyhan syndrome.
Catalog No. 50-004-13976 Supplier Medchemexpress LLC Supplier No. HYB1081A100MG
Oxidopamine (6-OHDA) hydrobromide is an antagonist of the neurotransmitter dopamine. It is a widely used neurotoxin that selectively destroys dopaminergic neurons. It promotes COX-2 activation, leading to PGE2 synthesis and pro-inflammatory cytokine IL-1β secretion. Oxidopamine hydrobromide can be used for the research of Parkinson's disease (PD), attention-deficit hyperactivity disorder (ADHD), and Lesch-Nyhan syndrome.
  • Antagonist of the neurotransmitter dopamine.
  • Widely used neurotoxin.
  • Selectively destroys dopaminergic neurons.
  • Promotes COX-2 activation.
  • Leads to PGE2 synthesis and pro-inflammatory cytokine IL-1β secretion.
  • Used for research of Parkinson's disease (PD), attention-deficit hyperactivity disorder (ADHD), and Lesch-Nyhan syndrome.