Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oxindole, 97%, pure

Catalog No. AC129670050
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle

Catalog No. AC129670050

Supplier: Thermo Scientific Chemicals 129670050

Only null left
Add to Cart

CAS: 59-48-3 | C8H7NO | 133.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59-48-3
Molecular Formula C8H7NO
Molecular Weight (g/mol) 133.15
MDL Number MFCD00005711
InChI Key JYGFTBXVXVMTGB-UHFFFAOYSA-N
Synonym oxindole, indolin-2-one, 2-oxindole, 2-indolinone, 1,3-dihydro-2h-indol-2-one, oxindol, 2-oxoindoline, 2-oxindoline, indol-2 3h-one, 2h-indol-2-one, 1,3-dihydro
PubChem CID 321710
ChEBI CHEBI:31697
IUPAC Name 1,3-dihydroindol-2-one
SMILES C1C2=CC=CC=C2NC1=O

Specifications

Melting Point 123°C to 128°C
Boiling Point 227°C (73.0 mmHg)
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Beilstein 21, 282
Solubility Information Solubility in water: insoluble. Other solubilities: soluble inn most organic solvents
Formula Weight 133.15
Percent Purity 97%
Grade Pure
Chemical Name or Material Oxindole, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 200-429-5
RTECSNumber NM2080500

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.